2-(Cyclobutylamino)nicotinic acid ≥97%

Supplier: Matrix Scientific

082486-1G
102837-442EA 949.23 USD
102837-442
2-(Cyclobutylamino)nicotinic acid ≥97%
2-(Cyclobutylamino)nicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MDL Number: MFCD16747178
CAS Number: 1343611-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR