Pyridin-2-ylacetic acid, lithium salt ≥97%

Supplier: Matrix Scientific

082480-1G 082480-5G
102837-430EA 1535.38 USD
102837-430 102837-428
Pyridin-2-ylacetic acid, lithium salt ≥97%
Pyridin-2-ylacetic acid, lithium salt

Hazard: Irritant

Formula: C₇H₆LiNO₂
MDL Number: MFCD23701769
CAS Number: 58197-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR