3-Amino-5-bromoisonicotinic acid ≥97%

Supplier: Matrix Scientific

082474-1G
102837-412EA 1118.46 USD
102837-412
3-Amino-5-bromoisonicotinic acid ≥97%
3-Amino-5-bromoisonicotinic acid

Hazard: Irritant

Formula: C₆H₅BrN₂O₂
MDL Number: MFCD18258608
CAS Number: 1256806-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR