6-(Cyclopent-3-en-1-yloxy)pyridine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082472-1G
102837-408EA 949.23 USD
102837-408
6-(Cyclopent-3-en-1-yloxy)pyridine-2-carboxylic acid ≥97%
6-(Cyclopent-3-en-1-yloxy)pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MDL Number: MFCD19706048
CAS Number: 1355334-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR