6-(Cyclopent-3-en-1-yloxy)nicotinic acid ≥97%

Supplier: Matrix Scientific

082471-1G
102837-406EA 949.23 USD
102837-406
6-(Cyclopent-3-en-1-yloxy)nicotinic acid ≥97%
6-(Cyclopent-3-en-1-yloxy)nicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MDL Number: MFCD19706044
CAS Number: 1355334-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR