2-(Cyclopent-3-en-1-yloxy)isonicotinic acid ≥97%

Supplier: Matrix Scientific

082470-1G
102837-404EA 949.23 USD
102837-404
2-(Cyclopent-3-en-1-yloxy)isonicotinic acid ≥97%
2-(Cyclopent-3-en-1-yloxy)isonicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MDL Number: MFCD19706028
CAS Number: 1355334-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR