6-(1,3-Dioxan-5-yloxy)nicotinic acid ≥97%

Supplier: Matrix Scientific

082463-1G
102837-388EA 863.08 USD
102837-388
6-(1,3-Dioxan-5-yloxy)nicotinic acid ≥97%
6-(1,3-Dioxan-5-yloxy)nicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₁NO₅
MDL Number: MFCD18089210
CAS Number: 1287217-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR