2-(Cyclopropylmethoxy)isonicotinic acid ≥97%
Supplier: Matrix Scientific
082460-1G
102837-382EA
863.08
USD
102837-382
2-(Cyclopropylmethoxy)isonicotinic acid ≥97%
2-(Cyclopropylmethoxy)isonicotinic acid
Hazard: Irritant
Formula:
C₁₀H₁₁NO₃ |
MDL Number:
MFCD11136674 CAS Number: 1019546-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...