4-(Furan-3-yl)pyridine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082398-1G
102837-256EA 949.23 USD
102837-256
4-(Furan-3-yl)pyridine-2-carboxylic acid ≥97%
4-(Furan-3-yl)pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₃
MDL Number: MFCD08056302
CAS Number: 914637-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR