6-[4-(Dimethylamino)butoxy]nicotinic acid ≥97%

Supplier: Matrix Scientific

082396-1G
102837-252EA 750.77 USD
102837-252
6-[4-(Dimethylamino)butoxy]nicotinic acid ≥97%
6-[4-(Dimethylamino)butoxy]nicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₃
MDL Number: MFCD14666473
CAS Number: 1287218-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR