2-[(Cyclohexylmethyl)amino]nicotinic acid ≥97%

Supplier: Matrix Scientific

082395-1G
102837-250EA 750.77 USD
102837-250
2-[(Cyclohexylmethyl)amino]nicotinic acid ≥97%
2-[(Cyclohexylmethyl)amino]nicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD12408728
CAS Number: 939819-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR