2-(Cyclobutylmethoxy)nicotinic acid ≥97%

Supplier: Matrix Scientific

082394-1G
102837-248EA 949.23 USD
102837-248
2-(Cyclobutylmethoxy)nicotinic acid ≥97%
2-(Cyclobutylmethoxy)nicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD13195607
CAS Number: 1235439-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR