6-[(Cyclohexylmethyl)amino]pyridine-2carboxylic acid ≥97%

Supplier: Matrix Scientific

082393-1G
102837-246EA 750.77 USD
102837-246
6-[(Cyclohexylmethyl)amino]pyridine-2carboxylic acid ≥97%
6-[(Cyclohexylmethyl)amino]pyridine-2carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MW: 234.3 g/mol
MDL Number: MFCD13195606
CAS Number: 1235440-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR