6-(Cyclobutylmethoxy)pyridine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082392-1G
102837-244EA 750.77 USD
102837-244
6-(Cyclobutylmethoxy)pyridine-2-carboxylic acid ≥97%
6-(Cyclobutylmethoxy)pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD13195605
CAS Number: 1235441-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR