6-(Cyclobutylmethoxy)nicotinic acid ≥97%

Supplier: Matrix Scientific

082390-1G
102837-240EA 750.77 USD
102837-240
6-(Cyclobutylmethoxy)nicotinic acid ≥97%
6-(Cyclobutylmethoxy)nicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD13195604
CAS Number: 1235440-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR