2-[(Cyclohexylmethyl)amino]isonicotinic acid ≥97%
Supplier: Matrix Scientific
082389-1G
102837-238EA
750.77
USD
102837-238
2-[(Cyclohexylmethyl)amino]isonicotinic acid ≥97%
2-[(Cyclohexylmethyl)amino]isonicotinic acid
Hazard: Irritant
Formula:
C₁₃H₁₈N₂O₂ |
MDL Number:
MFCD12408880 CAS Number: 1096303-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...