Difluoro(6-methylpyridin-2-yl)acetic acid ≥97%

Supplier: Matrix Scientific

082379-1G
102837-216EA 946.15 USD
102837-216
Difluoro(6-methylpyridin-2-yl)acetic acid ≥97%
Difluoro(6-methylpyridin-2-yl)acetic acid

Hazard: Irritant

Formula: C₈H₇F₂NO₂
MDL Number: MFCD12827869
CAS Number: 1215353-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR