Difluoro[3-(trifluoromethyl)pyridin-2-yl]acetic acid ≥97%

Supplier: Matrix Scientific

082376-1G
102837-210EA 652.31 USD
102837-210
Difluoro[3-(trifluoromethyl)pyridin-2-yl]acetic acid ≥97%
Difluoro[3-(trifluoromethyl)pyridin-2-yl]acetic acid

Hazard: Irritant

Formula: C₈H₄F₅NO₂
MDL Number: MFCD12963754
CAS Number: 1215579-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR