Difluoro[5-(trifluoromethyl)pyridin-2-yl]acetic acid ≥97%
Supplier: Matrix Scientific
082375-1G
102837-208EA
1461.54
USD
102837-208
Difluoro[5-(trifluoromethyl)pyridin-2-yl]acetic acid ≥97%
Difluoro[5-(trifluoromethyl)pyridin-2-yl]acetic acid
Hazard: Irritant
Formula:
C₈H₄F₅NO₂ |
MDL Number:
MFCD12827865 CAS Number: 1215583-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...