6-(2-Aziridin-1-ylethoxy)-5-bromonicotinic acid ≥97%

Supplier: Matrix Scientific

082354-1G
102837-152EA 863.08 USD
102837-152
6-(2-Aziridin-1-ylethoxy)-5-bromonicotinic acid ≥97%
6-(2-Aziridin-1-ylethoxy)-5-bromonicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₁BrN₂O₃
MDL Number: MFCD11100072
CAS Number: 1216703-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR