5-Bromo-6-(cyclohexyloxy)nicotinic acid ≥97%
Supplier: Matrix Scientific
082351-1G
102837-146EA
863.08
USD
102837-146
5-Bromo-6-(cyclohexyloxy)nicotinic acid ≥97%
5-Bromo-6-(cyclohexyloxy)nicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₄BrNO₃ |
MDL Number:
MFCD11100069 CAS Number: 1216394-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...