5-Bromo-6-(piperidin-4-yloxy)nicotinic acid ≥97%
Supplier: Matrix Scientific
082350-1G
102837-144EA
863.08
USD
102837-144
5-Bromo-6-(piperidin-4-yloxy)nicotinic acid ≥97%
5-Bromo-6-(piperidin-4-yloxy)nicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₃BrN₂O₃ |
MDL Number:
MFCD11100068 CAS Number: 1216671-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...