5-Bromo-6-(tetrahydro-2H-pyran-4-yloxy)nicotinic acid ≥97%

Supplier: Matrix Scientific

082349-1G
102837-142EA 863.08 USD
102837-142
5-Bromo-6-(tetrahydro-2H-pyran-4-yloxy)nicotinic acid ≥97%
5-Bromo-6-(tetrahydro-2H-pyran-4-yloxy)nicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₂BrNO₄
MDL Number: MFCD11100067
CAS Number: 1216775-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR