5-Bromo-6-(1H-imidazol-1-yl)nicotinic acid ≥97%

Supplier: Matrix Scientific

082348-1G
102837-140EA 863.08 USD
102837-140
5-Bromo-6-(1H-imidazol-1-yl)nicotinic acid ≥97%
5-Bromo-6-(1H-imidazol-1-yl)nicotinic acid

Hazard: Irritant

Formula: C₉H₆BrN₃O₂
MDL Number: MFCD11100066
CAS Number: 1216625-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR