5-Bromo-6-piperidin-1-ylnicotinic acid ≥97%
Supplier: Matrix Scientific
082346-1G
102837-136EA
863.08
USD
102837-136
5-Bromo-6-piperidin-1-ylnicotinic acid ≥97%
5-Bromo-6-piperidin-1-ylnicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₃BrN₂O₂ |
MDL Number:
MFCD11100064 CAS Number: 1216952-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...