5-Bromo-2-phenoxynicotinic acid ≥97%

Supplier: Matrix Scientific

082342-1G
102837-128EA 800 USD
102837-128
5-Bromo-2-phenoxynicotinic acid ≥97%
5-Bromo-2-phenoxynicotinic acid

Hazard: Irritant

Formula: C₁₂H₈BrNO₃
MDL Number: MFCD11100060
CAS Number: 1215394-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR