5-Bromo-2-(piperidin-4-yloxy)nicotinic acid ≥97%
Supplier: Matrix Scientific
082340-1G
102837-124EA
806.15
USD
102837-124
5-Bromo-2-(piperidin-4-yloxy)nicotinic acid ≥97%
5-Bromo-2-(piperidin-4-yloxy)nicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₃BrN₂O₃ |
MDL Number:
MFCD11100058 CAS Number: 1219201-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...