2-(Piperidin-4-ylmethoxy)nicotinic acid ≥97%
Supplier: Matrix Scientific
082335-1G
102837-114EA
636.92
USD
102837-114
2-(Piperidin-4-ylmethoxy)nicotinic acid ≥97%
2-(Piperidin-4-ylmethoxy)nicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₃ |
MDL Number:
MFCD11100053 CAS Number: 1208087-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...