2-(Tetrahydro-2H-pyran-4-yloxy)nicotinic acid ≥97%

Supplier: Matrix Scientific

082331-1G
102837-106EA 524.62 USD
102837-106
2-(Tetrahydro-2H-pyran-4-yloxy)nicotinic acid ≥97%
2-(Tetrahydro-2H-pyran-4-yloxy)nicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD11100050
CAS Number: 1208088-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR