2-(Tetrahydro-2H-pyran-4-ylmethoxy)isonicotinic acid

Supplier: Matrix Scientific

082317-1G
102837-078EA 707.69 USD
102837-078
2-(Tetrahydro-2H-pyran-4-ylmethoxy)isonicotinic acid
2-(Tetrahydro-2H-pyran-4-ylmethoxy)isonicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₄
MDL Number: MFCD11100040
CAS Number: 1215846-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR