2-(Piperidin-4-ylmethoxy)isonicotinic acid ≥97%
Supplier: Matrix Scientific
082313-1G
102837-070EA
524.62
USD
102837-070
2-(Piperidin-4-ylmethoxy)isonicotinic acid ≥97%
2-(Piperidin-4-ylmethoxy)isonicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₃ |
MDL Number:
MFCD11100036 CAS Number: 1215707-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...