3-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Boc-amino)pyridine-2-carboxylic acid

082309-1G
102837-062EA 636.92 USD
102837-062
3-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid ≥97%
3-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₄
MW: 238.25 g/mol
MDL Number: MFCD02181061
CAS Number: 569687-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR