2-(Piperidin-4-yloxy)isonicotinic acid ≥97%

Supplier: Matrix Scientific

082291-1G
102837-026EA 553.85 USD
102837-026
2-(Piperidin-4-yloxy)isonicotinic acid ≥97%
2-(Piperidin-4-yloxy)isonicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₃
MDL Number: MFCD09258834
CAS Number: 1086379-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR