2-(Tetrahydro-2H-pyran-4-yloxy)isonicotinic acid ≥97%

Supplier: Matrix Scientific

082288-1G
102837-020EA 750.77 USD
102837-020
2-(Tetrahydro-2H-pyran-4-yloxy)isonicotinic acid ≥97%
2-(Tetrahydro-2H-pyran-4-yloxy)isonicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD09258832
CAS Number: 1086379-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR