2-(2-Aziridin-1-ylethoxy)isonicotinic acid ≥97%

Supplier: Matrix Scientific

082286-1G
102837-016EA 553.85 USD
102837-016
2-(2-Aziridin-1-ylethoxy)isonicotinic acid ≥97%
2-(2-Aziridin-1-ylethoxy)isonicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃
MDL Number: MFCD09258830
CAS Number: 1086379-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR