4-(Tetrahydro-2H-pyran-4-ylmethoxy)benzoic acid ≥97%
Supplier: Matrix Scientific
082260-1G
102836-946EA
636.92
USD
102836-946
4-(Tetrahydro-2H-pyran-4-ylmethoxy)benzoic acid ≥97%
4-(Tetrahydro-2H-pyran-4-ylmethoxy)benzoic acid
Hazard: Irritant
Formula:
C₁₃H₁₆O₄ |
MDL Number:
MFCD08669640 CAS Number: 1008773-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...