3-(2-Methylphenyl)oxirane-2-carboxylic acid ≥97%
Supplier: Matrix Scientific
082202-1G
102836-802EA
806.15
USD
102836-802
3-(2-Methylphenyl)oxirane-2-carboxylic acid ≥97%
3-(2-Methylphenyl)oxirane-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₀O₃ |
MDL Number:
MFCD18089205 CAS Number: 1287218-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...