3-(4-Chlorophenyl)oxirane-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082195-1G
102836-788EA 806.15 USD
102836-788
3-(4-Chlorophenyl)oxirane-2-carboxylic acid ≥97%
3-(4-Chlorophenyl)oxirane-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₇ClO₃
MDL Number: MFCD18089198
CAS Number: 114380-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR