3-(3-Chlorophenyl)oxirane-2-carboxylic acid ≥97%
Supplier: Matrix Scientific
082194-1G
102836-786EA
806.15
USD
102836-786
3-(3-Chlorophenyl)oxirane-2-carboxylic acid ≥97%
3-(3-Chlorophenyl)oxirane-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₇ClO₃ |
MDL Number:
MFCD18089197 CAS Number: 1287218-00-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...