4-(6-Chloropyrimidin-4-yl)benzoic acid ≥97%

Supplier: Matrix Scientific

082166-1G
102836-724EA 1118.46 USD
102836-724
4-(6-Chloropyrimidin-4-yl)benzoic acid ≥97%
4-(6-Chloropyrimidin-4-yl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₇ClN₂O₂
MDL Number: MFCD09861805
CAS Number: 750559-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR