3-(5-Bromo-2-furyl)benzoic acid ≥97%

Supplier: Matrix Scientific

082165-1G
102836-722EA 949.23 USD
102836-722
3-(5-Bromo-2-furyl)benzoic acid ≥97%
3-(5-Bromo-2-furyl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₇BrO₃
MDL Number: MFCD13195600
CAS Number: 1235440-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR