2-(2-Chloro-4-fluorophenyl)-2-methylpropanoic acid ≥97%

Supplier: Matrix Scientific

082147-1G
102836-676EA 1504.62 USD
102836-676
2-(2-Chloro-4-fluorophenyl)-2-methylpropanoic acid ≥97%
2-(2-Chloro-4-fluorophenyl)-2-methylpropanoic acid

Hazard: Irritant

Formula: C₁₀H₁₀ClFO₂
MDL Number: MFCD11036917
CAS Number: 920501-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR