4-[3-(4-Fluoroyphenyl)prop-2-enoyl]benzoic acid ≥97%

Supplier: Matrix Scientific

082145-1G
102836-672EA 723.08 USD
102836-672
4-[3-(4-Fluoroyphenyl)prop-2-enoyl]benzoic acid ≥97%
4-[3-(4-Fluoroyphenyl)prop-2-enoyl]benzoic acid

Hazard: Irritant

Formula: C₁₆H₁₁FO₃
MDL Number: MFCD12404893
CAS Number: 62557-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR