2-(Pyrrolidin-1-ylsulfonyl)benzoic acid ≥97%

Supplier: Matrix Scientific

082136-1G
102836-650EA 666.15 USD
102836-650
2-(Pyrrolidin-1-ylsulfonyl)benzoic acid ≥97%
2-(Pyrrolidin-1-ylsulfonyl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄S
MDL Number: MFCD09804452
CAS Number: 893615-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR