2-(2,5-Dioxopyrrolidin-1-yl)benzoic acid ≥97%

Supplier: Matrix Scientific

082130-1G
102836-638EA 750.77 USD
102836-638
2-(2,5-Dioxopyrrolidin-1-yl)benzoic acid ≥97%
2-(2,5-Dioxopyrrolidin-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₉NO₄
MDL Number: MFCD00022590
CAS Number: 80022-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR