4-(3,5-Dimethylisoxazol-4-yl)benzoic acid ≥97%

Supplier: Matrix Scientific

082125-1G
102836-626EA 461.54 USD
102836-626
4-(3,5-Dimethylisoxazol-4-yl)benzoic acid ≥97%
4-(3,5-Dimethylisoxazol-4-yl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MDL Number: MFCD12026348
CAS Number: 212515-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR