4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid ≥97%

Supplier: Matrix Scientific

082112-1G
102836-586EA 340 USD
102836-586
4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid ≥97%
4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzoic acid

Hazard: Irritant

Formula: C₁₃H₇ClF₃NO₃
MDL Number: MFCD11840267
CAS Number: 105626-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR