3-[(2-Chloropyrimidin-4-yl)amino]benzoic acid ≥97%

Supplier: Matrix Scientific

082107-1G
102836-576EA 636.92 USD
102836-576
3-[(2-Chloropyrimidin-4-yl)amino]benzoic acid ≥97%
3-[(2-Chloropyrimidin-4-yl)amino]benzoic acid

Hazard: Irritant

Formula: C₁₁H₈ClN₃O₂
MDL Number: MFCD03002171
CAS Number: 1208083-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR