2-((2,2-Dinitrilovinyl)amino)benzoic acid ≥95%

Supplier: Matrix Scientific

082091-1G
102836-528EA 666.15 USD
102836-528
2-((2,2-Dinitrilovinyl)amino)benzoic acid ≥95%
2-((2,2-Dinitrilovinyl)amino)benzoic acid

Hazard: Irritant

Formula: C₁₁H₇N₃O₂
MW: 213.2 g/mol
MDL Number: MFCD00170479
CAS Number: 811841-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR