4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid ≥97%

Supplier: Matrix Scientific

082054-1G
102836-432EA 496.92 USD
102836-432
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid ≥97%
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid

Hazard: Irritant

Formula: C₁₃H₈F₃NO₃
MDL Number: MFCD08448326
CAS Number: 869109-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR